Anti-BID + FAS Antibody Pair

Supplier: Abnova
ABNODI0181EA 802 CHF
ABNODI0181
Anti-BID + FAS Antibody Pair
Antibodies
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the BID protein, and the other against the FAS protein for use in Proximity Ligation Assay

Antibody pair set content: 1. BID Rabbit purified Polyclonal Antibody (20 µg) 2. FAS Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: BID + FAS
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR