Anti-HNF4A + CTNNB1 Antibody Pair

Supplier: Abnova
ABNODI0123EA 802 CHF
ABNODI0123
Anti-HNF4A + CTNNB1 Antibody Pair
Antibodies
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the HNF4A protein, and the other against the CTNNB1 protein for use in Proximity Ligation Assay

Antibody pair set content: 1. HNF4A Rabbit purified Polyclonal Antibody (20 µg) 2. CTNNB1 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: HNF4A + CTNNB1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR