Anti-RCVRN Polyclonal Antibody Pair
Supplier: Abnova
ABNOH00005957-AP21EA 1010 CHF
ABNOH00005957-AP21
Anti-RCVRN Polyclonal Antibody Pair
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human RCVRN.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-RCVRN (100 µg); 2. Detection Antibody: Mouse Polyclonal Anti-RCVRN (40 µl);
Type:
Antigen: RCVRN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human