Anti-CNDP2 Polyclonal Antibody Pair

Supplier: Abnova
ABNOH00055748-AP21EA 1010 CHF
ABNOH00055748-AP21
Anti-CNDP2 Polyclonal Antibody Pair
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human CNDP2.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-CNDP2 (100 µg); 2. Detection Antibody: Mouse Polyclonal Anti-CNDP2 (40 µl);

Type:
Antigen: CNDP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR