Anti-GRIN2B Antibody Pair

Supplier: Abnova
ABNODP0192EA 882 CHF
ABNODP0192
Anti-GRIN2B Antibody Pair
Antibodies
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the GRIN2B protein, and the other against the specific Y1472 phosphorylated site of GRIN2B for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-GRIN2B Y1472 Rabbit Polyclonal Antibody (20 µl) In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. GRIN2B Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: GRIN2B
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR