Anti-STAT5A + STAT5A Antibody Pair

Supplier: Abnova
ABNODP0117EA 882 CHF
ABNODP0117
Anti-STAT5A + STAT5A Antibody Pair
Antibodies
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the STAT5A protein, and the other against the specific Y694 phosphorylated site of STAT5A for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-STAT5A Y694 Rabbit purified Polyclonal Antibody (20 µl) With 0.09% sodium azide; 2. STAT5A Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: STAT5A + STAT5A
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR