Rapamycin (aus Streptomyces hygroscopicus)

Lieferant: ADIPOGEN

ADIPAGCN20025M001EA 45 CHF
ADIPAGCN20025M001
Rapamycin (aus Streptomyces hygroscopicus)
Rapamycin

Rapamycin, isolated from Streptomyces hygroscopicus, forms a complex with FKBP12 and inhibits the molecular target of rapamycin (mTOR). It blocks the activation of T and B cells.


  • Antibiotic. Antibacterial and antifungal properties
  • Potent immunosuppressant used as an alternative to calcineurin inhibitors
  • Cell cycle progression (at the G1/S transition) inhibitor
  • Antiproliferative, antitumor compound, anti-HIV and anti-aging compound
  • Apoptosis enhancer and activator of autophagy
  • Purity: ≥98%


Stability: Soluble in DMSO, methanol or chloroform. Stable for at least 2 years after receipt when stored at long term storage −20 °C. Short term storage at 4 °C.

Formel: C₅₁H₇₉NO₁₃
MW: 914,19 g/mol
MDL Number: MFCD00867594
CAS-Nummer: 53123-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR