Anti-TAS2R20 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25086EA 622 CHF
ABNOPAB25086
Anti-TAS2R20 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of TAS2R49.

  • Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: TAS2R20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR