Anti-UMOD Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25083EA 622 CHF
ABNOPAB25083
Anti-UMOD Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of UMOD.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: UMOD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR