Anti-OR5W2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25080EA 545 CHF
ABNOPAB25080
Anti-OR5W2 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of OR5W2.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: OR5W2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR