Anti-POT1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25079EA 622 CHF
ABNOPAB25079
Anti-POT1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of POT1.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: POT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR