Anti-CALCA Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25076EA 622 CHF
ABNOPAB25076
Anti-CALCA Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of CALCA.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: CALCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR