Anti-KLHL13 Mouse Monoclonal Antibody [clone: 8D1]

Lieferant: Abnova
ABNOMAB10720EA 622 CHF
ABNOMAB10720
Anti-KLHL13 Mouse Monoclonal Antibody [clone: 8D1]
Antikörper
Mouse monoclonal antibody raised against partial recombinant KLHL13.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: KLHL13
Clonality: Monoclonal
Clone: 8D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR