Anti-KIT Mouse Monoclonal Antibody [clone: 1C5]

Lieferant: Abnova
ABNOMAB10712EA 622 CHF
ABNOMAB10712
Anti-KIT Mouse Monoclonal Antibody [clone: 1C5]
Antikörper
Mouse monoclonal antibody raised against partial recombinant KIT.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000

Type: Primary
Antigen: KIT
Clonality: Monoclonal
Clone: 1C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR