Anti-CCDC114 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB23951EA 622 CHF
ABNOPAB23951
Anti-CCDC114 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant CCDC114.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000

Type: Primary
Antigen: CCDC114
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR