Anti-CEACAM5 Mouse Monoclonal Antibody [clone: CB30]

Lieferant: Abnova
ABNOMAB3848EA 479 CHF
ABNOMAB3848
Anti-CEACAM5 Mouse Monoclonal Antibody [clone: CB30]
Antikörper
Mouse monoclonal antibody raised against CEACAM5.

Recommended Dilutions: Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 10 ?g/ml; Immunoprecipitation

Type: Primary
Antigen: CEACAM5
Clonality: Monoclonal
Clone: CB30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR