Anti-C-peptide Mouse Monoclonal Antibody [clone: C-PEP-01]

Lieferant: Abnova
ABNOMAB3849EA 479 CHF
ABNOMAB3849
Anti-C-peptide Mouse Monoclonal Antibody [clone: C-PEP-01]
Antikörper
Mouse monoclonal antibody raised against synthetic peptide of C-peptide.

Type: Primary
Antigen: C-peptide
Clonality: Monoclonal
Clone: C-PEP-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR