Anti-immunodeficiency virus protease Mouse Monoclonal Antibody [clone: 1696]

Lieferant: Abnova
ABNOMAB3853EA 479 CHF
ABNOMAB3853
Anti-immunodeficiency virus protease Mouse Monoclonal Antibody [clone: 1696]
Antikörper
Mouse monoclonal antibody raised against full length recombinant human immunodeficiency virus protease.

Recommended Dilutions: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: immunodeficiency virus protease
Clonality: Monoclonal
Clone: 1696
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR