Anti-HLA-G Mouse Monoclonal Antibody [clone: MEM-G/9]

Lieferant: Abnova
ABNOMAB3855EA 428 CHF
ABNOMAB3855
Anti-HLA-G Mouse Monoclonal Antibody [clone: MEM-G/9]
Antikörper
Mouse monoclonal antibody raised against recombinant HLA-G.

Recommended Dilutions: ELISA: 10 ?g/ml; Flow Cytometry: 1-5 ?g/ml; Immunocytochemistry Immunohistochemistry: Formalin/PFA-fixed paraffin-embedded sections; Immunoprecipitation

Type: Primary
Antigen: HLA-G
Clonality: Monoclonal
Clone: MEM-G/9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR