Anti-PRDM14 Rabbit Polyclonal Antibody

ABNOPAB2396EA 422 CHF
ABNOPAB2396
Anti-PRDM14 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of PRDM14.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500; Immunohistochemistry: 1:50-100

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500; Immunohistochemistry: 1:50-100

Type: Primary
Antigen: PRDM14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR