Anti-MAP2 Mouse Monoclonal Antibody [clone: MT-01]

Lieferant: Abnova
ABNOMAB3866EA 479 CHF
ABNOMAB3866
Anti-MAP2 Mouse Monoclonal Antibody [clone: MT-01]
Antikörper
Mouse monoclonal antibody raised against full length native MAP2.

Type: Primary
Antigen: MAP2
Clonality: Monoclonal
Clone: MT-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Cat, Human, Mouse, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR