Anti-C19ORF70 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB23966EA 622 CHF
ABNOPAB23966
Anti-C19ORF70 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant C19orf70.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20

Type: Primary
Antigen: C19ORF70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR