Anti-TTLL10 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB23970EA 654 CHF
ABNOPAB23970
Anti-TTLL10 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant TTLL10.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: TTLL10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR