Anti-SRFBP1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB23973EA 622 CHF
ABNOPAB23973
Anti-SRFBP1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant SRFBP1.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2500; Western Blot: 1:250-1:500

Type: Primary
Antigen: SRFBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR