Anti-C1QTNF6 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB13158EA 622 CHF
ABNOPAB13158
Anti-C1QTNF6 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of C1QTNF6.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: C1QTNF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR