Anti-IRAK2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB13161EA 622 CHF
ABNOPAB13161
Anti-IRAK2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of Irak2.

Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: IRAK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR