Anti-GPR17 Rabbit Polyclonal Antibody
CAYM10136-1EA 623 CHF
CAYM10136-1
Anti-GPR17 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GPR17
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat