Anti-MAGE-1 Mouse Monoclonal Antibody [clone: MA454]

Lieferant: United States Biological
USBI138566EA 493 CHF
USBI138566
Anti-MAGE-1 Mouse Monoclonal Antibody [clone: MA454]
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml; ; Immunofluorescence: 1-2?g/ml; Immunoprecipitation: 1-2?g/500 ?g; Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: MAGE-1
Clonality: Monoclonal
Clone: MA454
Conjugation: unconjugated
Epitope: full-length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Dog, Human, Rat,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR