Anti-Keratin 18 Mouse Monoclonal Antibody [clone: DC10]

Lieferant: United States Biological
USBI138559-20EA 474 CHF
USBI138559-20
Anti-Keratin 18 Mouse Monoclonal Antibody [clone: DC10]
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml; ; Immunofluorescence: 1-2?g/ml; Immunoprecipitation: 1-2?g/500 ?g; Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: Keratin 18
Clonality: Monoclonal
Clone: DC10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR