Anti-Keratin 18 Mouse Monoclonal Antibody [clone: DA7]

Lieferant: United States Biological
USBI138558-20EA 474 CHF
USBI138558-20
Anti-Keratin 18 Mouse Monoclonal Antibody [clone: DA7]
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml; ; Immunofluorescence: 1-2?g/ml; Immunoprecipitation: 1-2?g/500 ?g; Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: Keratin 18
Clonality: Monoclonal
Clone: DA7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR