Anti-Granulocyte Mouse Monoclonal Antibody [clone: BM-2]

Lieferant: United States Biological
USBI138554-20EA 474 CHF
USBI138554-20
Anti-Granulocyte Mouse Monoclonal Antibody [clone: BM-2]
Antikörper
Recommended dilution: ; Immunofluorescence: 1-2?g/ml; Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: Granulocyte
Clonality: Monoclonal
Clone: BM-2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Macaque,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR