Anti-Estrogen Receptor, beta Mouse Monoclonal Antibody [clone: ERb455]

Lieferant: United States Biological
USBI138550-20EA 474 CHF
USBI138550-20
Anti-Estrogen Receptor, beta Mouse Monoclonal Antibody [clone: ERb455]
Antikörper
Recommended dilution: Immunofluorescence: 1-2?g/ml; Western Blot: 0.5-1 ?g/ml; Immunochemistry: 0.5-1 ?g/ml; Immunoprecipitation: 1-2?g/500 ?g

Type: Primary
Antigen: Estrogen Receptor, beta
Clonality: Monoclonal
Clone: ERb455
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Horse, Human, Monkey, Mouse, Pig, Rat, Sheep,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR