Anti-EpCAM Mouse Monoclonal Antibody [clone: MOC-31]

Lieferant: United States Biological
USBI138544-100EA 380 CHF
USBI138544-100
Anti-EpCAM Mouse Monoclonal Antibody [clone: MOC-31]
Antikörper
Recommended dilution: Immunofluorescence: 1:50-1:100; Western Blot: 1:100-1:200; Immunoprecipitation: 5-10?l/500 ?g; Immunochemistry: 1:100-1:200

Type: Primary
Antigen: EpCAM
Clonality: Monoclonal
Clone: MOC-31
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR