Anti-Cytokeratin, pan Mouse Monoclonal Antibody [clone: AE-1/AE-3]

Lieferant: United States Biological
USBI138538-20EA 474 CHF
USBI138538-20
Anti-Cytokeratin, pan Mouse Monoclonal Antibody [clone: AE-1/AE-3]
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunofluorescence: 1-2?g/ml; Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: Cytokeratin, pan
Clonality: Monoclonal
Clone: AE-1/AE-3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Bovine, Dog, Chicken, Human, Monkey, Mouse, Rabbit, Rat,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR