Anti-CD8a Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI138530-100EA 813 CHF
USBI138530-100 USBI138530
Anti-CD8a Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 1-2?g/ml; Immunoprecipitation: 1-2?g/500 ?g; Western Blot: 1-2?g/ml; Immunochemistry: 1-2?g/ml

Type: Primary
Antigen: CD8a
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR