Anti-CD56 Mouse Monoclonal Antibody [clone: 123C3.D5]
Lieferant: United States Biological
USBI138521-100EA 813 CHF
USBI138521-100 USBI138521
Anti-CD56 Mouse Monoclonal Antibody [clone: 123C3.D5]
Antikörper
Recommended dilution: ; Immunofluorescence: 1-2?g/ml; Western Blot: 0.5-1 ?g/ml; Immunoprecipitation: 1-2?g/500 ?g; Immunochemistry: 0.5-1 ?g/ml
Type: Primary
Antigen: CD56
Clonality: Monoclonal
Clone: 123C3.D5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat, Zebrafish,