Anti-CD5 Mouse Monoclonal Antibody [clone: C5/473]

Lieferant: United States Biological
USBI138520EA 380 CHF
USBI138520
Anti-CD5 Mouse Monoclonal Antibody [clone: C5/473]
Antikörper
Recommended dilution: Immunofluorescence: 1:50-1:100; Western Blot: 1:100-1:200; Immunoprecipitation: 5-10?l/500 ?g; Immunochemistry: 1:100-1:200

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: C5/473
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR