Anti-HGF Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: United States Biological
USBIH2005-08A-APEA 1200 CHF
USBIH2005-08A-AP
Anti-HGF Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: HGF (hepatocyte growth factor (hepapoietin A; scatter factor))
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human