Anti-Hepatitis C Virus Core Mouse Monoclonal Antibody [clone: 8.V.023]

Lieferant: United States Biological
USBIH1925-21-100EA 0 CHF
USBIH1925-21-100
Anti-Hepatitis C Virus Core Mouse Monoclonal Antibody [clone: 8.V.023]
Antikörper
Type: Primary
Antigen: Hepatitis C Virus Core
Clonality: monoclonal
Clone: 8.V.023
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR