Anti-Hepatitis C Virus Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIH1924-80EEA 896 CHF
USBIH1924-80E
Anti-Hepatitis C Virus Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: Hepatitis C Virus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR