Anti-Hepatitis C Virus NS5 Mouse Monoclonal Antibody [clone: 10D110]
Lieferant: United States Biological
USBIH1920-27T2EA 863 CHF
USBIH1920-27T2
Anti-Hepatitis C Virus NS5 Mouse Monoclonal Antibody [clone: 10D110]
Antikörper
Type: Primary
Antigen: Hepatitis C Virus NS5
Clonality: monoclonal
Clone: 10D110
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now