Anti-CST7 Goat polyclonal antibody

Lieferant: United States Biological
USBIC8950-23EA 698 CHF
USBIC8950-23
Anti-CST7 Goat polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:8000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: CST7 (cystatin F (leukocystatin))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR