Anti-SLAMF8 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIB2106-02AEA 632 CHF
USBIB2106-02A
Anti-SLAMF8 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-4 ?g/ml

Type: Primary
Antigen: SLAMF8 (SLAM family member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR