Anti-LAD1 Mouse Monoclonal Antibody [clone: 2A9]

Lieferant: United States Biological
USBI128984EA 733 CHF
USBI128984
Anti-LAD1 Mouse Monoclonal Antibody [clone: 2A9]
Antikörper
Type: Primary
Antigen: LAD1 (ladinin 1)
Clonality: monoclonal
Clone: 2A9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR