Anti-Lactate Dehydrogenase Rabbit Polyclonal Antibody
Lieferant: United States Biological
USBI128983EA 788 CHF
USBI128983
Anti-Lactate Dehydrogenase Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Lactate Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human