Anti-L3MBTL4 Mouse Monoclonal Antibody [clone: 1B9-1C12]

Supplier: US Biological
USBI128980EA 702 CHF
USBI128980
Anti-L3MBTL4 Mouse Monoclonal Antibody [clone: 1B9-1C12]
Antibodies
Type: Primary
Antigen: L3MBTL4 (l(3)mbt-like 4 (Drosophila))
Clonality: monoclonal
Clone: 1B9-1C12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR