Anti-L3MBTL4 Rabbit Polyclonal Antibody

Supplier: US Biological
USBI128979EA 750 CHF
USBI128979
Anti-L3MBTL4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: L3MBTL4 (l(3)mbt-like 4 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR