Anti-L3MBTL2 Mouse Monoclonal Antibody [clone: 4F4]
Lieferant: United States Biological
USBI128977EA 733 CHF
USBI128977
Anti-L3MBTL2 Mouse Monoclonal Antibody [clone: 4F4]
Antikörper
Type: Primary
Antigen: L3MBTL2 (l(3)mbt-like 2 (Drosophila))
Clonality: monoclonal
Clone: 4F4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human