Anti-KYNU Mouse Monoclonal Antibody [clone: 1G2]

Lieferant: United States Biological
USBI128975EA 733 CHF
USBI128975
Anti-KYNU Mouse Monoclonal Antibody [clone: 1G2]
Antikörper
Type: Primary
Antigen: KYNU (kynureninase)
Clonality: monoclonal
Clone: 1G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR