Anti-KSR1 Mouse Monoclonal Antibody [clone: 6C7]

Supplier: US Biological
USBI128971EA 702 CHF
USBI128971
Anti-KSR1 Mouse Monoclonal Antibody [clone: 6C7]
Antibodies
Type: Primary
Antigen: KSR1 (kinase suppressor of ras 1)
Clonality: monoclonal
Clone: 6C7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR